Imprimer

Anti-SLC22A13 Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT3908AEA 350 EUR
ABGEAT3908A
Anti-SLC22A13 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: SLC22A13 (solute carrier family 22 (organic anion/urate transporter), member 13)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR