Anti-SLC25A13 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT3911AEA 350 EUR
ABGEAT3911A
Anti-SLC25A13 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: SLC25A13 (solute carrier family 25 (aspartate/glutamate carrier), member 13)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR