Anti-SLC26A2 Mouse Monoclonal Antibody
Supplier: Abgent
ABGEAT3914AEA 350 EUR
ABGEAT3914A
Anti-SLC26A2 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: SLC26A2 (solute carrier family 26 (anion exchanger), member 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human