Anti-SLC33A1 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT3921AEA 350 EUR
ABGEAT3921A
Anti-SLC33A1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: SLC33A1 (solute carrier family 33 (acetyl-CoA transporter), member 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR