Anti-SLC4A4 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT3927AEA 350 EUR
ABGEAT3927A
Anti-SLC4A4 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: SLC4A4 (solute carrier family 4 (sodium bicarbonate cotransporter), member 4)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR