Anti-SLC4A8 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT3928AEA 350 EUR
ABGEAT3928A
Anti-SLC4A8 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: SLC4A8 (solute carrier family 4, sodium bicarbonate cotransporter, member 8)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR