Anti-SMAD1 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT3936AEA 350 EUR
ABGEAT3936A
Anti-SMAD1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: SMAD1 (SMAD family member 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR