Anti-SMU1 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT3965AEA 350 EUR
ABGEAT3965A
Anti-SMU1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: SMU1 (smu-1 suppressor of mec-8 and unc-52 homolog (C. elegans))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR