Imprimer

Anti-SNAPC5 Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT3977AEA 350 EUR
ABGEAT3977A
Anti-SNAPC5 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: SNAPC5 (small nuclear RNA activating complex, polypeptide 5, 19kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR