Imprimer
Anti-SNRPA Mouse Monoclonal Antibody
Fournisseur: Abgent
ABGEAT3982AEA 350 EUR
ABGEAT3982A
Anti-SNRPA Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: SNRPA (small nuclear ribonucleoprotein polypeptide A)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human