Imprimer

Anti-SOCS3 Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT3988AEA 350 EUR
ABGEAT3988A
Anti-SOCS3 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: SOCS3 (suppressor of cytokine signaling 3)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR