Imprimer

Anti-SOX15 Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT3995AEA 350 EUR
ABGEAT3995A
Anti-SOX15 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: SOX15 (SRY (sex determining region Y)-box 15)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR