Imprimer

Anti-SPDEF Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT4018AEA 350 EUR
ABGEAT4018A
Anti-SPDEF Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: SPDEF (SAM pointed domain containing ETS transcription factor)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR