Imprimer

Anti-SRCRB4D Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT4034AEA 350 EUR
ABGEAT4034A
Anti-SRCRB4D Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: SRCRB4D (scavenger receptor cysteine rich domain containing, group B (4 domains))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR