Anti-SREBF1 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT4035AEA 350 EUR
ABGEAT4035A
Anti-SREBF1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: SREBF1 (sterol regulatory element binding transcription factor 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR