Imprimer

Anti-SRPK1 Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT4041AEA 350 EUR
ABGEAT4041A
Anti-SRPK1 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: SRPK1 (SRSF protein kinase 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR