Anti-ST6GAL1 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT4052AEA 350 EUR
ABGEAT4052A
Anti-ST6GAL1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: ST6GAL1 (ST6 beta-galactosamide alpha-2,6-sialyltranferase 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR