Imprimer

Anti-STAMBP Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT4058AEA 350 EUR
ABGEAT4058A
Anti-STAMBP Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: STAMBP (STAM binding protein)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR