Imprimer

Anti-STAU2 Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT4071AEA 350 EUR
ABGEAT4071A
Anti-STAU2 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: STAU2 (staufen double-stranded RNA binding protein 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR