Anti-STEAP1 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT4074AEA 350 EUR
ABGEAT4074A
Anti-STEAP1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: STEAP1 (six transmembrane epithelial antigen of the prostate 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR