Imprimer

Anti-SUGT1 Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT4103AEA 350 EUR
ABGEAT4103A
Anti-SUGT1 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: SUGT1 (SGT1, suppressor of G2 allele of SKP1 (S. cerevisiae))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR