Imprimer

Anti-SUPT16H Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT4106AEA 350 EUR
ABGEAT4106A
Anti-SUPT16H Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: SUPT16H (suppressor of Ty 16 homolog (S. cerevisiae))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR