Anti-SUPT3H Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT4107AEA 350 EUR
ABGEAT4107A
Anti-SUPT3H Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: SUPT3H (suppressor of Ty 3 homolog (S. cerevisiae))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR