Imprimer

Anti-TAF12 Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT4142AEA 350 EUR
ABGEAT4142A
Anti-TAF12 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: TAF12 (TAF12 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 20kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR