Imprimer

Anti-TAF1C Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT4144AEA 350 EUR
ABGEAT4144A
Anti-TAF1C Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: TAF1C (TATA box binding protein (TBP)-associated factor, RNA polymerase I, C, 110kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR