Imprimer

Anti-TAF1L Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT4145AEA 350 EUR
ABGEAT4145A
Anti-TAF1L Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: TAF1L (TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 210kDa-like)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR