Anti-TAF5L Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT4146AEA 350 EUR
ABGEAT4146A
Anti-TAF5L Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: TAF5L (TAF5-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associated factor, 65kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR