Anti-TAF7 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT4148AEA 350 EUR
ABGEAT4148A
Anti-TAF7 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: TAF7 (TAF7 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 55kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR