Imprimer

Anti-TAX1BP1 Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT4156AEA 350 EUR
ABGEAT4156A
Anti-TAX1BP1 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: TAX1BP1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR