Anti-TAX1BP3 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT4158AEA 350 EUR
ABGEAT4158A
Anti-TAX1BP3 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: TAX1BP3
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR