Anti-TBX21 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT4167AEA 350 EUR
ABGEAT4167A
Anti-TBX21 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: TBX21 (T-box 21)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR