Imprimer

Anti-TBX5 Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT4171AEA 350 EUR
ABGEAT4171A
Anti-TBX5 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: TBX5 (T-box 5)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR