Imprimer

Anti-TCEB2 Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT4179AEA 350 EUR
ABGEAT4179A
Anti-TCEB2 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: TCEB2 (transcription elongation factor B (SIII), polypeptide 2 (18kDa, elongin B))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR