Imprimer
Anti-TCF7L2 Mouse Monoclonal Antibody
Fournisseur: Abgent
ABGEAT4191AEA 350 EUR
ABGEAT4191A
Anti-TCF7L2 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: TCF7L2 (transcription factor 7-like 2 (T-cell specific, HMG-box))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human