Imprimer

Anti-TCOF1 Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT4197AEA 350 EUR
ABGEAT4197A
Anti-TCOF1 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: TCOF1 (Treacher Collins-Franceschetti syndrome 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR