Anti-TFAP2B Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT4212AEA 350 EUR
ABGEAT4212A
Anti-TFAP2B Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: TFAP2B (transcription factor AP-2 beta (activating enhancer binding protein 2 beta))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR