Anti-TFAP4 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT4214AEA 350 EUR
ABGEAT4214A
Anti-TFAP4 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: TFAP4 (transcription factor AP-4 (activating enhancer binding protein 4))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR