Anti-TFF3 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT4224AEA 350 EUR
ABGEAT4224A
Anti-TFF3 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: TFF3 (trefoil factor 3 (intestinal))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR