Anti-TFPI2 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT4225AEA 350 EUR
ABGEAT4225A
Anti-TFPI2 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: TFPI2 (tissue factor pathway inhibitor 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR