Imprimer

Anti-THAP1 Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT4231AEA 350 EUR
ABGEAT4231A
Anti-THAP1 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: THAP1 (THAP domain containing, apoptosis associated protein 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR