Imprimer

Anti-TIMM8A Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT4239AEA 350 EUR
ABGEAT4239A
Anti-TIMM8A Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: TIMM8A (translocase of inner mitochondrial membrane 8 homolog A (yeast))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR