Imprimer

Anti-TP63 Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT4314AEA 350 EUR
ABGEAT4314A
Anti-TP63 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: TP63 (tumor protein p63)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR