Imprimer

Anti-TPSAB1 Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT4326AEA 350 EUR
ABGEAT4326A
Anti-TPSAB1 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: TPSAB1 (tryptase alpha/beta 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR