Imprimer

Anti-TRAF3IP2 Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT4330AEA 350 EUR
ABGEAT4330A
Anti-TRAF3IP2 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: TRAF3IP2 (TRAF3 interacting protein 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR