Imprimer
Anti-TRPA1 Mouse Monoclonal Antibody
Fournisseur: Abgent
ABGEAT4367AEA 350 EUR
ABGEAT4367A
Anti-TRPA1 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: TRPA1 (transient receptor potential cation channel, subfamily A, member 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human