Anti-TRPC4AP Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT4368AEA 350 EUR
ABGEAT4368A
Anti-TRPC4AP Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: TRPC4AP (transient receptor potential cation channel, subfamily C, member 4 associated protein)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR