Anti-TUBA1C Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT4401AEA 350 EUR
ABGEAT4401A
Anti-TUBA1C Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: TUBA1C (tubulin, alpha 1c)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR