Imprimer

Anti-TWIST2 Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT4412AEA 350 EUR
ABGEAT4412A
Anti-TWIST2 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: TWIST2 (twist family bHLH transcription factor 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR