Imprimer

Anti-ERP44 Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT4417AEA 350 EUR
ABGEAT4417A
Anti-ERP44 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: ERP44 (endoplasmic reticulum protein 44)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR