Anti-PNPLA2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA301537EA 569 EUR
ORIGTA301537
Anti-PNPLA2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: PNPLA2 (patatin-like phospholipase domain containing 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR